Order Entry
Ireland
ContactUsLinkComponent
Anti-MUS81 Mouse Monoclonal Antibody [clone: MTA30 2G10/3]
Anti-MUS81 Mouse Monoclonal Antibody [clone: MTA30 2G10/3]
  ABCAAB14387-100
 :  Abcam
Anti-MUS81 Mouse Monoclonal Antibody [clone: MTA30 2G10/3]
  ABCAAB14387-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    MUS81 structure-specific endonuclease subunit
  • Antigen symbol:
    MUS81
  • Clonality:
    Monoclonal
  • Clone:
    MTA30 2G10/3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MUS81 (MUS81 structure-specific endonuclease subunit)
Clonality: monoclonal
Clone: MTA30 2G10/3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human