Order Entry
Ireland
ContactUsLinkComponent
Anti-POU1F1 Mouse Monoclonal Antibody [clone: 5E4]
Anti-POU1F1 Mouse Monoclonal Antibody [clone: 5E4]
Catalog # ABCAAB10545-500
Supplier:  Abcam
Anti-POU1F1 Mouse Monoclonal Antibody [clone: 5E4]
Catalog # ABCAAB10545-500
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    POU class 1 homeobox 1
  • Antigen symbol:
    POU1F1
  • Clonality:
    Monoclonal
  • Clone:
    5E4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: POU1F1 (POU class 1 homeobox 1)
Clonality: monoclonal
Clone: 50000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Mouse