Order Entry
Ireland
ContactUsLinkComponent
Anti-Reticulon 1A Mouse Monoclonal Antibody [clone: MON162]
Anti-Reticulon 1A Mouse Monoclonal Antibody [clone: MON162]
Catalog # ABCAAB9274-100
Supplier:  Abcam
undefined
Anti-Reticulon 1A Mouse Monoclonal Antibody [clone: MON162]
Catalog # ABCAAB9274-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Reticulon 1A
  • Clonality:
    Monoclonal
  • Clone:
    MON162
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MON162] to Reticulon 1A.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Reticulon 1A
Clonality: Monoclonal
Clone: MON162
Conjugation:
Epitope: The epitope of this antibody is located between amino acid residues 338-422 of Reticulon-1A
Host: Mouse
Isotype: IgG1
Reactivity: Human;Pig