Anti-Hepatitis C Virus Core 1b Mouse Monoclonal Antibody [clone: C7-50]
Catalog # ABCAAB2740-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis C Virus Core Antigen
- Clonality:Monoclonal
- Clone:C7-50
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pack type:Vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C7-50] to Hepatitis C Virus Core 1b.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Hepatitis C Virus Core 1b
Clonality: Monoclonal
Clone: C7-50
Conjugation:
Epitope: This antibody recognizes an epitope between amino acid residues 21-40 of HCV core protein. This sequence is conserved among different HCV strains.
Host: Mouse
Isotype: IgG1
Reactivity: HCV