Anti-ADAM22 Mouse Monoclonal Antibody
BIRBORB99081-100
: Biorbyt
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 22
- Antigen symbol:ADAM22
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Amino acid number:444 to 526
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Fusion protein amino acids 444-526 (extracellulardisintegrin domain) of mouseADAM22, accession numberQ9R1V6
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1 ?g/ml
Type: Primary
Antigen: ADAM22 (ADAM metallopeptidase domain 22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat