Anti-Ig Pig Polyclonal Antibody
Catalog # BIRBORB98176-10
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin
- Antigen symbol:Ig
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Pig
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Pig
Isotype: IgG
Reactivity: Human