Anti-Crotalus durissus terrificus venom Phosphodiesterase I Rabbit Polyclonal Antibody (Biotin)
BIRBORB98145-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Crotalus durissus terrificus venom Phosphodiesterase I
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Snake
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Crotalus durissus terrificus venom Phosphodiesterase I
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Snake