Anti-PDZD8 Rabbit Polyclonal Antibody
Catalog # BIRBORB96127-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:PDZ Domain Containing 8
- Antigen symbol:PDZD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Storage buffer:PBS, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: PDZD8 (PDZ domain containing 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human