Anti-LZTS2 Rabbit Polyclonal Antibody
BIRBORB89898-50
: Biorbyt
- Antibody type:Primary
- Antigen name:leucine zipper, putative tumor suppressor 2
- Antigen symbol:LZTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: LZTS2 (leucine zipper, putative tumor suppressor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human