Anti-MAP2K1 Rabbit Polyclonal Antibody
Catalog # BIRBORB88794-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 1
- Antigen symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, pH 7,2, 50% glycerol, 0,09% azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: