Anti-ENaC Gamma Rabbit Polyclonal Antibody
BIRBORB88314-50
: Biorbyt
- Antibody type:Primary
- Antigen name:ENAC gamma
- Antigen symbol:ENACG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 5 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: ENaC Gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human