Order Entry
Ireland
ContactUsLinkComponent
Anti-HTR3A Rabbit Polyclonal Antibody
Anti-HTR3A Rabbit Polyclonal Antibody
  BIRBORB87928-50
 :  Biorbyt
undefined
Anti-HTR3A Rabbit Polyclonal Antibody
  BIRBORB87928-50
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    5-hydroxytryptamine (serotonin) receptor 3A, ionotropic
  • Antigen symbol:
    HTR3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage buffer:
    PBS, 0,2% gelatin, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Immunochemistry: 5 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: HTR3A (5-hydroxytryptamine (serotonin) receptor 3A, ionotropic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: