Anti-Phosphodiesterase 6a Rabbit Polyclonal Antibody
Catalog # BIRBORB86313-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Phosphodiesterase 6a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic 18 amino acid peptide from N-terminus of human PDE6A/PDE6 Alpha
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 10-15 ?g/ml
Type: Primary
Antigen: Phosphodiesterase 6a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human