Anti-OAS1 Rabbit Polyclonal Antibody
BIRBORB78226-100
: Biorbyt
- Antibody type:Primary
- Antigen name:2'-5'-oligoadenylate synthetase 2, 69/71kDa
- Antigen symbol:OAS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Recombinant protein
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:20000-1:80000; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: OAS1 (2'-5'-oligoadenylate synthetase 1, 40/46kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human