Anti-neutrophil CAP18 Mouse Monoclonal Antibody
Catalog # BIRBORB76596-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:neutrophil CAP18
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage buffer:Purified antibody containing PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Partially purified human neutrophil membrane proteins heparin Ultrogellectin affinity purification fractions in octyl glucoside
- Purification:purified by Protein G chromatography
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: neutrophil CAP18
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human