Anti-COX6A2 Rabbit Polyclonal Antibody
Catalog # BIRBORB69731-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cytochrome c oxidase subunit VIa polypeptide 2
- Antigen symbol:COX6A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:0,9 mg/ml
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: COX6A2 (cytochrome c oxidase subunit VIa polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human