Anti-RSF1 Rabbit Polyclonal Antibody
Catalog # BIRBORB67050-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Remodeling and spacing factor 1
- Antigen symbol:RSF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C-terminal region of human RSF1
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Western Blot: 1:1000-1:5000
Type: Primary
Antigen: RSF1 (remodeling and spacing factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human