Anti-ABCG1 Rabbit Polyclonal Antibody
BIRBORB67027-100
: Biorbyt
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family G, member 1
- Antigen symbol:ABCG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Middle region of human ABCG1
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Western Blot: 1:1000-1:5000
Type: Primary
Antigen: ABCG1 (ATP-binding cassette, sub-family G (WHITE), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human