Anti-Caspase-3 (P10) Rabbit Polyclonal Antibody
BIRBORB137850-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Caspase 3 (p10)
- Antigen symbol:CASP3 (p10)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminal of mouse Caspase-3 (P10), different from the related rat sequence by one amino acid.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Caspase-3 (P10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat