Anti-RAC2 Mouse Monoclonal Antibody
BIRBORB137443-50
: Biorbyt
- Antibody type:Primary
- Antigen name:RAS-related C3 botulinum substrate 2
- Antigen symbol:RAC2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:RAC2 antibody was raised against recombinant human RAC2 (1-189aa) protein purified from E. coli.
- Purification:purified by Protein G chromatography
- Size:50 µl
- Pk:50 µl
Immunochemistry: 10 ?g/ml; Western Blot: 1:1000
Type: Primary
Antigen: RAC2 (ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human