Anti-SEPT2 Rabbit Polyclonal Antibody
: Biorbyt
Type: Primary
Antigen: SEPT2 (septin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
- Catalog No:
- BIRBORB127607-200
- BIRBORB127607-100
- Antigen symbol:
- MARCH10
- MARCH7
- Antigen name:
- Membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase
- Membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase
- Antigen synonyms:
- RNF190|MARCH-X
- AXOT|RNF177|AXO|MARCH-VII
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together