Anti-MAP3K5 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BIRBORB122989-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase kinase 5
- Antigen symbol:MAP3K5
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Thr845
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:155 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human ASK1 around the phosphorylation site of Thr845.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MAP3K5 (mitogen-activated protein kinase kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: