Anti-DNA Topoisomerase 1 Rabbit Polyclonal Antibody
Catalog # BIRBORB109559-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:DNA Topoisomerase 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide made to human DNA Topoisomerase 1
- Purification:purified by antigen immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DNA Topoisomerase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human