Anti-25(OH)Vitamin D3-1-alpha Hydroxylase Sheep Polyclonal Antibody
BIRBORB27523-100
: Biorbyt
- Antibody type:Primary
- Antigen name:25-Hydroxy-Vitamin D3-1-alpha Hydroxylase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Format:concentrated
- Form:liquid
- Amino acid number:266 to 289
- Storage temperature:Cool
- Concentration:1/500 (depending on fixation, species, unmasking method and secondary system)
- Shipping temperature:Ice
- Immunogen:Synthetic peptide from the coding sequence of murine 25(OH)-Vitamin-D3-1-alpha-Hydroxylase (aa 266-289)
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: 25(OH)Vitamin D3-1-alpha Hydroxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: