Anti-CML (N-Epsilon)-Carboxymethyl-Lysine Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB27501-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:N-Epsilon-Carboxymethyl-Lysine
- Antigen symbol:CML
- Clonality:Monoclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Format:concentrated
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:N-Epsilon-(Carboxymethyl)-Lysine (CML)-HSA
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml
Type: Primary
Antigen: CML (N-Epsilon)-Carboxymethyl-Lysine
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: