Anti-COL4 Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
ImmunofluorescenceA 1:80; Immunochemistry: 1:500 - 1:1000
Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB27318-0.1
- BIRBORB27318-0.5
- Antigen symbol:
- COL4
- COL4
- Antigen name:
- collagen, type IV
- collagen, type IV
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Purified collagen type IV from human placenta
- Purified collagen type IV from human placenta
- Form:
- lyophilized
- lyophilized
- Purification:
- purified
- purified
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together