Anti-COL3 Rabbit Polyclonal Antibody
BIRBORB26846-0.5
: Biorbyt
- Antibody type:Primary
- Antigen name:collagen, type III
- Antigen symbol:COL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Collagen type III extracted and purified from Human Placenta.
- Purification:purified by chromatography
- Size:500 μL
- Pk:0,5 mL
Immunofluorescence: 1:80; Immunochemistry: 1:500; ELISA: 1:200
Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human