Anti-Peroxidase (HRP) Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB25366-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Horseradish Peroxidase
- Antigen symbol:HRP
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Sheep
- Form:lyophilized
- Storage buffer:0,01M Sodium Phosphate, 0,14M Sodium Chloride, pH 7,4
- Storage temperature:Cool
- Concentration:10 mg/ml (OD280) (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Peroxidase (Horseradish)
- Purification:purified by delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:50 mg
- Pk:50 mg
Specifications
About this item
Type: Primary
Antigen: Peroxidase (HRP)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: