Anti-Herpes simplex Type-2 Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB23097-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Herpes simplex Type-2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Sheep
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage buffer:PBS containing 10 mg/mL BSA
- Storage temperature:Cool
- Concentration:1-2 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:HSV Type 2 Strain G (Human)
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: Herpes simplex Type-2
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: