Anti-IgA1/IgA2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB22224-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgA1/IgA2
- Clonality:Monoclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- Isotype:IgG1
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:0,4 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) approximately 1,2, No foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Highly purified monoclonal IgA isolated from human serum.
- Size:200 μg
- Pk:0,2 mg
Specifications
About this item
Type: Secondary
Antigen: IgA1+IgA2
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: