Anti-kappa light chain Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanante))
Catalog # BIRBORB21963-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Goat
- Format:immunoconjugate
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein ratio (F/P) approximately 1,9, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:A pool of purified Bence Jones kappa proteins isolated from human urine. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Goat
Isotype: none
Reactivity: