Anti-IgG1/IgG2a/IgG2b/IgG3 Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BIRBORB21667-1
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG3
- Clonality:Polyclonal
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,8, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG, including all known subclasses, isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: none
Reactivity: