Anti-IgG1/IgG2a/IgG2b/IgG2c Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21479-2
Supplier: Biorbyt
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG1/IgG2a/IgG2b/IgG2c
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- Format:immunoconjugate
- Form:lyophilized
- Storage temperature:Cool
- Concentration:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,7, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Purified normal IgG including all known subclasses isolated from pooled rat serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:2 ml
- Pk:2 mL
Specifications
About this item
Type: Secondary
Antigen: Rat IgG1 IgG2A IgG2B IgG2C
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: