Anti-Polynucleotide Phosphorylase Rabbit Polyclonal Antibody (Biotin)
BIRBORB21039-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Polynucleotide Phosphorylase
- Antigen symbol:Polynucleotide Phosphorylase
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Bacteria
- Format:immunoconjugate
- Storage buffer:phosphate buffered saline
- Storage temperature:Cool
- Concentration:1 mg/ml, Biotin/IgG protein molar ratio (B/P) approximately 4,3, no foreign proteins added,
- Shipping temperature:Ice
- Immunogen:Polynuleotide phosphorylase isolated and purified from Micrococcus luteus. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: Polynucleotide Phosphorylase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bacteria