Anti-NF200 Mouse Monoclonal Antibody
Catalog # BIRBORB18247-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Neurofilament, Heavy Polypeptide
- Antigen symbol:NEFH
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized
- Storage buffer:1.2% sodium acetate or neutral PBS
- Storage temperature:Cool
- Concentration:100 ug/ml
- Shipping temperature:Ice
- Immunogen:C-terminal segment of enzymatically dephosphorylated pig Neurofilament 200.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NF200
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human, Mouse, Rat