Anti-ATF2 Rabbit polyclonal antibody
Catalog # BIRBORB14763-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 55
- Antigen symbol:ATF55
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:ATF2
- Modification:phospho-Ser62 or 44
- Storage buffer:PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human ATF2 around the phosphorylation site of serine 62 or 44 (N -D-SP-V-I).
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Immunochemistry: 1:50-1:100
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat