Anti-SLC27A1 Mouse Monoclonal Antibody [clone: 2G9]
ABNOH00376497-M03
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 27 Member A1
- Antigen symbol:Solute carrier family 27, member 1
- Clonality:Monoclonal
- Clone:2G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:376497
- Antigen synonyms:hFACVL1|VLACS|VLCS|FACVL1|FATP2|HsT17226|ACSVL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SLC27A1
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human