Anti-OTOP2 Mouse Monoclonal Antibody [clone: 4F6]
ABNOH00092736-M10
: Abnova
- Antibody type:Primary
- Antigen name:Otopetrin 2
- Antigen symbol:OTOP2
- Clonality:Monoclonal
- Clone:4F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:92736
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: OTOP2
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human