Anti-LNX1 Mouse Monoclonal Antibody [clone: 4E3]
Catalog # ABNOH00084708-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ligand of numb-protein X 1
- Antigen symbol:LNX1
- Clonality:Monoclonal
- Clone:4E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:84708
- Antigen synonyms:MPDZ|LNX|PDZRN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LNX1
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human