Anti-HINT2 Mouse Monoclonal Antibody [clone: 4E11]
Catalog # ABNOH00084681-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:histidine triad nucleotide binding protein 2
- Antigen symbol:HINT2
- Clonality:Monoclonal
- Clone:4E11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:84681
- Antigen synonyms:HIT-17
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HINT2
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human