Anti-TRIM63 Rabbit Polyclonal Antibody
Catalog # ABNOH00084676-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 63
- Antigen symbol:TRIM63
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:84676
- Antigen synonyms:tripartite motif-containing 63|muscle specific ring finger protein 2|MURF1|IRF|tripartite motif-containing protein 63|SMRZ|RNF28|iris ring finger protein|striated muscle RING zinc finger protein|MURF2|E3 ubiquitin-protein ligase TRIM63|muscle-specific RING finger protein 1|ring finger protein 28
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TRIM63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse