Anti-IGDCC4 Mouse Monoclonal Antibody [clone: 6E6]
Catalog # ABNOH00057722-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin Superfamily, DCC Subclass, Member 4
- Antigen symbol:IGDCC4
- Clonality:Monoclonal
- Clone:6E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:57722
- Antigen synonyms:DDM36|Nope
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: IGDCC4
Clonality: Monoclonal
Clone: 6E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human