Anti-PCDHA2 Mouse Monoclonal Antibody [clone: 2G12]
Catalog # ABNOH00056146-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protocadherin alpha 2
- Antigen symbol:PCDHA2
- Clonality:Monoclonal
- Clone:2G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:56146
- Antigen synonyms:PCDH-ALPHA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PCDHA2
Clonality: Monoclonal
Clone: 2G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human