Anti-CFC1 Rabbit Polyclonal Antibody
ABNOH00055997-D01
: Abnova
- Antibody type:Primary
- Antigen name:Cripto, FRL-1, cryptic family 1
- Antigen symbol:CFC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:55997
- Antigen synonyms:HTX2|CFC1B|CRYPTIC|DTGA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: CFC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human