Anti-DTL Rabbit Polyclonal Antibody
Catalog # ABNOH00051514-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:denticleless E3 ubiquitin protein ligase homolog (Drosophila)
- Antigen symbol:DTL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51514
- Antigen synonyms:L2DTL|DCAF2|CDT2|RAMP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DTL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human