Anti-EHD3 Mouse Polyclonal Antibody
ABNOH00030845-A01
: Abnova
- Antibody type:Primary
- Antigen name:EH-domain containing 3
- Antigen symbol:EHD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:30845
- Antigen synonyms:PAST3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: EHD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat