Anti-HEY1 Mouse Monoclonal Antibody [clone: 1E10]
ABNOH00023462-M05
: Abnova
- Antibody type:Primary
- Antigen name:Hairy/enhancer-of-split related with YRPW motif 1
- Antigen symbol:HEY1
- Clonality:Monoclonal
- Clone:1E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23462
- Antigen synonyms:MGC1274|HESR1|BHLHb31|OAF1|HERP2|CHF2|HRT-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: HEY1
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human