Anti-DZIP1 Mouse Polyclonal Antibody
Catalog # ABNOH00022873-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DAZ interacting zinc finger protein 1
- Antigen symbol:DZIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:22873
- Antigen synonyms:DZIPt1|Zinc finger DAZ interacting protein 1|DZIP|Zinc finger protein DZIP1|DAZ interacting protein testis1|DZIP2|Zinc finger protein DZIPt1.|DAZ interacting protein 1/2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DZIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human