Anti-CDK2AP2 Mouse Polyclonal Antibody
ABNOH00010263-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 2 associated protein 2
- Antigen symbol:CDK2AP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10263
- Antigen synonyms:DOC-1R|p14
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CDK2AP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat