Order Entry
Ireland
ContactUsLinkComponent
Anti-COL4A3BP Mouse Monoclonal Antibody [clone: 1A1]
Anti-COL4A3BP Mouse Monoclonal Antibody [clone: 1A1]
Catalog # ABNOH00010087-M02
Supplier:  Abnova
undefined
Anti-COL4A3BP Mouse Monoclonal Antibody [clone: 1A1]
Catalog # ABNOH00010087-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
  • Antigen symbol:
    COL4A3BP
  • Clonality:
    Monoclonal
  • Clone:
    1A1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    10087
  • Antigen synonyms:
    STARD11|CERT|CERTL|GPBP|MRD34
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: COL4A3BP
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human